Learn More
Abnova™ Human CHMP5 Full-length ORF (NP_057494.2, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051510-P01.25ug
Additional Details : Weight : 0.00010kg
Description
CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A (MIM 164010), is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 [PubMed 16730941]).[supplied by OMIM]
Sequence: MNRLFGKAKPKAPPPSLTGCIGTVDSRAESIDKKISRLDAELVKYKDQIKKMREGPAKNMVKQKALRVLKQKRMYEQQRDNLAQQSFNMEQANYTIQSLKDTKTTVDAMKLGVKEMKKAYKQVKIDQIEDLQDQLEDMMEDANEIQEALSRSYGTPELDEDDLEAELDALGDELLADEDSSYLDEAASAPAIPEGVPTDTKNKDGVLVDEFGLPQIPASSpecifications
NP_057494.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C9orf83/CGI-34/HSPC177/PNAS-2/SNF7DC2 | |
CHMP5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51510 | |
CHMP5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNRLFGKAKPKAPPPSLTGCIGTVDSRAESIDKKISRLDAELVKYKDQIKKMREGPAKNMVKQKALRVLKQKRMYEQQRDNLAQQSFNMEQANYTIQSLKDTKTTVDAMKLGVKEMKKAYKQVKIDQIEDLQDQLEDMMEDANEIQEALSRSYGTPELDEDDLEAELDALGDELLADEDSSYLDEAASAPAIPEGVPTDTKNKDGVLVDEFGLPQIPAS | |
RUO | |
CHMP5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |