missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CEP112 Full-length ORF (NP_001032402.1, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00201134-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139263

  • 508.00 € / 25µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein with filament, myosin tail and ATPase domains. Orthologs of this gene exist in mouse, rat and chimp. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MWASLSLDHPSAKENQALRLIEMREENGNVPKTEQAGSLKPLRDTGKSNLKEKKANSKLKQIEKEYTQKLAKSSQIIAELQTTISSLKEENSQQQLAAERRLQDVRQKFEDEKKQLIRDNDQAIKVLQDELENRSNQVRCAEKKLQHKELESQEQITYIRQEYETKLKGLMPASLRQELEDTISSLKSQVNFLQKRASILQEELTTYQGRR
Specifications

Specifications

NP_001032402.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCDC46/MACOCO
CEP112
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
201134
CEP112 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWASLSLDHPSAKENQALRLIEMREENGNVPKTEQAGSLKPLRDTGKSNLKEKKANSKLKQIEKEYTQKLAKSSQIIAELQTTISSLKEENSQQQLAAERRLQDVRQKFEDEKKQLIRDNDQAIKVLQDELENRSNQVRCAEKKLQHKELESQEQITYIRQEYETKLKGLMPASLRQELEDTISSLKSQVNFLQKRASILQEELTTYQGRR
RUO
CEP112
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions