Learn More
Abnova™ Human CAPN7 Partial ORF (NP_055111.1, 714 a.a. - 813 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_055111.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23473 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16174726
|
Abnova™
H00023473-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16184726
|
Abnova™
H00023473-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Calpains are ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. The calpain proteins are heterodimers consisting of an invariant small subunit and variable large subunits. The large subunit possesses a cysteine protease domain, and both subunits possess calcium-binding domains. Calpains have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. The function of the protein encoded by this gene is not known. An orthologue has been found in mouse but it seems to diverge from other family members. The mouse orthologue is thought to be calcium independent with protease activity. [provided by RefSeq]
Sequence: NPIYQFHIEKTGPLLIELRGPRQYSVGFEVVTVSTLGDPGPHGFLRKSSGDYRCGFCYLELENIPSGIFNIIPSTFLPKQEGPFFLDFNSIIPIKITQLQSpecifications
NP_055111.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CALPAIN7/FLJ36423/PALBH | |
CAPN7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23473 | |
CAPN7 (Human) Recombinant Protein (Q01) | |
NPIYQFHIEKTGPLLIELRGPRQYSVGFEVVTVSTLGDPGPHGFLRKSSGDYRCGFCYLELENIPSGIFNIIPSTFLPKQEGPFFLDFNSIIPIKITQLQ | |
RUO | |
CAPN7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |