Learn More
Abnova™ Human CACNG7 Full-length ORF (NP_114102.2, 1 a.a. - 275 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00059284-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
The mouse protein stargazin is one of five subunits comprising neuronal voltage-gated calcium channels. This subunit, gamma, is thought to stabilize the calcium channel in an inactive (closed) state. Mutations in the gene encoding stargazin have been associated with absence seizures, also known as petit-mal or spike-wave seizures. The protein encoded by this gene is structurally similar to the mouse stargazin protein and is a member of the neuronal calcium channel gamma subunit protein family. However, it appears unlikely that the encoded protein is part of a functional calcium channel. Rather, it appears to inhibit the expression of a specific calcium channel subunit. [provided by RefSeq]
Sequence: MSHCSSRALTLLSSVFGACGLLLVGIAVSTDYWLYMEEGTVLPQNQTTEVKMALHAGLWRVCFFAGREKGRCVASEYFLEPEINLVTENTENILKTVRTATPFPMVSLFLVFTAFVISNIGHIRPQRTILAFVSGIFFILSGLSLVVGLVLYISSINDEVMNRPSSSEQYFHYRYGWSFAFAASSFLLKEGAGVMSVYLFTKRYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSPCSpezifikation
NP_114102.2 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CACNG7 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
59284 | |
CACNG7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSHCSSRALTLLSSVFGACGLLLVGIAVSTDYWLYMEEGTVLPQNQTTEVKMALHAGLWRVCFFAGREKGRCVASEYFLEPEINLVTENTENILKTVRTATPFPMVSLFLVFTAFVISNIGHIRPQRTILAFVSGIFFILSGLSLVVGLVLYISSINDEVMNRPSSSEQYFHYRYGWSFAFAASSFLLKEGAGVMSVYLFTKRYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSPC | |
RUO | |
CACNG7 | |
Yes |