Learn More
Abnova™ Human CACNG7 Full-length ORF (NP_114102.2, 1 a.a. - 275 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_114102.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 59284 |
Molecular Weight (g/mol) | 57.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193143
|
Abnova™
H00059284-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183143
|
Abnova™
H00059284-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The mouse protein stargazin is one of five subunits comprising neuronal voltage-gated calcium channels. This subunit, gamma, is thought to stabilize the calcium channel in an inactive (closed) state. Mutations in the gene encoding stargazin have been associated with absence seizures, also known as petit-mal or spike-wave seizures. The protein encoded by this gene is structurally similar to the mouse stargazin protein and is a member of the neuronal calcium channel gamma subunit protein family. However, it appears unlikely that the encoded protein is part of a functional calcium channel. Rather, it appears to inhibit the expression of a specific calcium channel subunit. [provided by RefSeq]
Sequence: MSHCSSRALTLLSSVFGACGLLLVGIAVSTDYWLYMEEGTVLPQNQTTEVKMALHAGLWRVCFFAGREKGRCVASEYFLEPEINLVTENTENILKTVRTATPFPMVSLFLVFTAFVISNIGHIRPQRTILAFVSGIFFILSGLSLVVGLVLYISSINDEVMNRPSSSEQYFHYRYGWSFAFAASSFLLKEGAGVMSVYLFTKRYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSPCSpecifications
NP_114102.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSHCSSRALTLLSSVFGACGLLLVGIAVSTDYWLYMEEGTVLPQNQTTEVKMALHAGLWRVCFFAGREKGRCVASEYFLEPEINLVTENTENILKTVRTATPFPMVSLFLVFTAFVISNIGHIRPQRTILAFVSGIFFILSGLSLVVGLVLYISSINDEVMNRPSSSEQYFHYRYGWSFAFAASSFLLKEGAGVMSVYLFTKRYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSPC | |
RUO | |
CACNG7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
59284 | |
CACNG7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CACNG7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |