Learn More
Abnova™ Human C17orf38 Partial ORF (NP_001010855, 661 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00146850-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Phosphoinositide 3-kinase gamma is a lipid kinase that produces the lipid second messenger phosphatidylinositol 3,4,5-trisphosphate. The kinase is composed of a catalytic subunit and one of several regulatory subunits, and is chiefly activated by G protein-coupled receptors. This gene encodes a regulatory subunit, and is distantly related to the phosphoinositide-3-kinase, regulatory subunit 5 gene which is located adjacent to this gene on chromosome 7. The orthologous protein in the mouse binds to both the catalytic subunit and to G(beta/gamma), and mediates activation of the kinase subunit downstream of G protein-coupled receptors. [provided by RefSeq]
Sequence: GKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQSpecifications
NP_001010855 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQ | |
RUO | |
PIK3R6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
146850 | |
C17orf38 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C17orf38/DKFZp666P158/FLJ34500/HsT41028/p84/p87(PIKAP)/p87PIKAP | |
PIK3R6 | |
Recombinant | |
wheat germ expression system |