missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AXIN1 Partial ORF (NP_003493, 643 a.a. - 740 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008312-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16084985

  • 518.16 € / 25µg

Estimated Shipment: 23-12-2021
Add to basket

Description

Description

This gene encodes a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a dishevelled and axin (DIX) domain. The encoded protein interacts with adenomatosis polyposis coli, catenin (cadherin-associated protein), beta 1, 88kDa, glycogen synthase kinase 3 beta, protein phosphate 2, and itself. This protein functions as a negative regulator of the wingless-type MMTV integration site family, member 1 (WNT) signaling pathway and can induce apoptosis. The crystal structure of a portion of this protein, alone and in a complex with other proteins, has been resolved. Mutations in this gene have been associated with hepatocellular carcinoma, hepatoblastomas, ovarian endometriod adenocarcinomas, and medullablastomas. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: ISRHRRTGHGSSGTRKPQPHENSRPLSLEHPWAGPQLRTSVQPSHLFIQDPTMPPHPAPNPLTQLEEARRRLEEEEKRASRAPSKQRYVQEVMRRGRA
Specifications

Specifications

NP_003493
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ISRHRRTGHGSSGTRKPQPHENSRPLSLEHPWAGPQLRTSVQPSHLFIQDPTMPPHPAPNPLTQLEEARRRLEEEEKRASRAPSKQRYVQEVMRRGRA
RUO
AXIN1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8312
AXIN1 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AXIN/MGC52315
AXIN1
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions