missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASNSD1 Full-length ORF (NP_061921.1, 1 a.a. - 643 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054529-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100623

  • 518.16 € / 25µg

Estimated Shipment: 03-01-2022
Add to basket

Description

Description

Sequence: MCGICCSVNFSAEHFSQDLKEDLLYNLKQRGPNSSKQLLKSDVNYQCLFSAHVLHLRGVLTTQPVEDERGNVFLWNGEIFSGIKVEAEENDTQILFNYLSSCKNESEILSLFSEVQGPWSFIYYQASSHYLWFGRDFFGRRSLLWHFSNLGKSFCLSSVGTQTSGLANQWQEVPASGLFRIDLKSTVISRCIILQLYPWKYISRENIIEENVNSLSQISADLPAFVSVVANEAKLYLEKPVVPLNMMLPQAALETHCSNISNVPPTREILQVFLTDVHMKEVIQQFIDVLSVAVKKRVLCLPRDENLTANEVLKTCDRKANVAILFSGGIDSMVIATLADRHIPLDEPIDLLNVAFIAEEKTMPTTFNREGNKQKNKCEIPSEEFSKDVAAAAADSPNKHVSVPDRITGRAGLKELQAVSPSRIWNFVEINVSMEELQKLRRTRICHLIRPLDTVLDDSIGCAVWFASRGIGWLVAQEGVKSYQSNAKVVLTGIGADEQLAGYSRHRVRFQSHGLEGLNKEIMMELGRISSRNLGRDDRVIGDHGKEARFPFLDENVVSFLNSLPIWEKANLTLPRGIGEKLLLRLAAVELGLTASALLPKRAMQFGSRIAKMEKINEKASDKCGRLQIMSLENLSIEKETKL
Specifications

Specifications

NP_061921.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
98.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ASNSD1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
54529
ASNSD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20752/NBLA00058/NS3TP1
ASNSD1
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions