Learn More
Abnova™ Human ARHGEF7 Partial ORF (NP_663788, 102 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_663788 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8874 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16129515
|
Abnova™
H00008874-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139515
|
Abnova™
H00008874-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho GTPases play a fundamental role in numerous cellular processes triggered by extracellular stimuli that work through G protein coupled receptors. The encoded protein belongs to a family of cytoplasmic proteins that activate the Ras-like family of Rho proteins by exchanging bound GDP for GTP. It forms a complex with the small GTP binding protein Rac1 and recruits Rac1 to membrane ruffles and to focal adhesions. This protein can induce membrane ruffling. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: SSLVTLNKVTADIGLGSDSVCARPSSHRIKSFDSLGSQSLHTRTSKLFQGQYRSLDMTDNSNNQLVVRAKFNFQQTNEDELSFSKGDVISpecifications
NP_663788 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BETA-PIX/COOL1/DKFZp686C12170/DKFZp761K1021/KIAA0142/KIAA0412/Nbla10314/P50/P50BP/P85/P85COOL1/P85SPR/PAK3/PIXB | |
ARHGEF7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8874 | |
ARHGEF7 (Human) Recombinant Protein (Q01) | |
SSLVTLNKVTADIGLGSDSVCARPSSHRIKSFDSLGSQSLHTRTSKLFQGQYRSLDMTDNSNNQLVVRAKFNFQQTNEDELSFSKGDVI | |
RUO | |
ARHGEF7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |