missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF4 Partial ORF (NP_056135, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_056135 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 50649 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045485
|
Abnova™
H00050649-Q02.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035485
|
Abnova™
H00050649-Q02.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene. [provided by RefSeq]
Sequence: MPWEEPAGEKPSCSHSQKAFHMEPAQKPCFTTDMVTWALLCISAETVRGEAPSQPRGIPHRSPVSVDDLWLEKTQRKKLQKQAHVERRLHIGAVHKDGVKSpecifications
NP_056135 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASEF/ASEF1/GEF4/STM6 | |
ARHGEF4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
50649 | |
ARHGEF4 (Human) Recombinant Protein (Q02) | |
MPWEEPAGEKPSCSHSQKAFHMEPAQKPCFTTDMVTWALLCISAETVRGEAPSQPRGIPHRSPVSVDDLWLEKTQRKKLQKQAHVERRLHIGAVHKDGVK | |
RUO | |
ARHGEF4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |