missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AP1G2 Partial ORF (NP_003908, 686 a.a. - 785 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008906-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139625

  • 508.00 € / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is compsed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. This protein along with the complex is thought to function at some trafficking step in the complex pathways between the trans-Golgi network and the cell surface. There are two alternatively spliced transcript variants of this gene encoding the same protein. [provided by RefSeq]

Sequence: RPPENPALLLITITATNFSEGDVTHFICQAAVPKSLQLQLQAPSGNTVPARGGLPITQLFRILNPNKAPLRLKLRLTYDHFHQSVQEIFEVNNLPVESWQ
Specifications

Specifications

NP_003908
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RPPENPALLLITITATNFSEGDVTHFICQAAVPKSLQLQLQAPSGNTVPARGGLPITQLFRILNPNKAPLRLKLRLTYDHFHQSVQEIFEVNNLPVESWQ
RUO
AP1G2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8906
AP1G2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
G2AD
AP1G2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions