Learn More
Abnova™ Human AK3L1 Partial ORF (NP_982289, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_982289 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 205 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184484
|
Abnova™
H00000205-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174484
|
Abnova™
H00000205-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1. [provided by RefSeq]
Sequence: MASKLLRAVILGPPGSGKGTVCQRIAQNFGLQHLSSGHFLRENIKASTEVGEMAKQYIEKSLLVPDHVITRLMMSELENRRGQHWLLDGFPRTLGQAEALDKICEVDLVISpecifications
NP_982289 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AK3/AK4/MGC166959 | |
AK3L1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
205 | |
AK3L1 (Human) Recombinant Protein (Q01) | |
MASKLLRAVILGPPGSGKGTVCQRIAQNFGLQHLSSGHFLRENIKASTEVGEMAKQYIEKSLLVPDHVITRLMMSELENRRGQHWLLDGFPRTLGQAEALDKICEVDLVI | |
RUO | |
AK3L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |