Learn More
Abnova™ Human AHRR Partial ORF (NP_065782, 617 a.a. - 715 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_065782 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57491 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139126
|
Abnova™
H00057491-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149126
|
Abnova™
H00057491-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dioxin is a teratogen that exerts its effects through the arylhydrocarbon receptor in conjunction with the receptor's binding partner, arylhydrocarbon receptor nuclear translocator. The protein encoded by this gene represses signal transduction by the arylhydrocarbon receptor by competing with the arylhydrocarbon receptor nuclear translocator for binding to the arylhydrocarbon receptor. Expression of the repressor is stimulated by the receptor/translocator heterodimer, thereby regulating receptor function through a negative feedback mechanism. In addition, the encoded protein can bind to nuclear factor kappa-B. [provided by RefSeq]
Sequence: RATAGRSRELTPFHPAHCACLEPTDGLPQSEPPHQLCARGRGEQSCTCRAAEAAPVVKREPLDSPQWATHSQGMVPGMLPKSALATLVPPQASGCTFLPSpecifications
NP_065782 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AHH/AHHR/KIAA1234/MGC167813/MGC176630/bHLHe77 | |
AHRR | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57491 | |
AHRR (Human) Recombinant Protein (Q01) | |
RATAGRSRELTPFHPAHCACLEPTDGLPQSEPPHQLCARGRGEQSCTCRAAEAAPVVKREPLDSPQWATHSQGMVPGMLPKSALATLVPPQASGCTFLP | |
RUO | |
AHRR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |