missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AHI1 Full-length ORF (AAH94800.1, 1 a.a. - 609 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number AAH94800.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54806
Molecular Weight (g/mol) 96.4kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16180763
View Documents Promotion Details
Abnova™
H00054806-P01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16190763
View Documents Promotion Details
Abnova™
H00054806-P01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MPTAESEAKVKTKVRFEELLKTHSDLMREKKKLKKKLVRSEENISPDTIRSNLHYMKETTSDDPDTIRSNLPHIKETTSDDVSAANTNNLKKSTRVTKNKLRNTQLATENPNGDASVEEDKQGKPNKKVIKTVPQLTTQDLKPETPENKVDSTHQKTHTKPQPGVDHQKSEKANEGREETDLEEDEELMQAYQCHVTEEMAKEIKRKIRKKLKEQLTYFPSDTLFHDDKLSSEKRKKKKEVPVFSKAETSTLTISGDTVEGEQKKESSVRSVSSDSHQDDEISSMEQSTEDSMQDDTKPKPKKTKKKTKAVADNNEDVDGDGVHEITSRDSPVYPKCLLDDDLVLGVYIHRTDRLKSDFMISHPMVKIHVVDEHTGQYVKKDDSGRPVSSYYEKENVDYILPIMTQPYDFKQLKSRLPEWEEQIVFNENFPYLLRGSDESPKVILFFEILDFLSVDEIKNNSEVQNQECGFRKIAWAFLKLLGANGNANINSKLRLQLYYPPTKPRSPLSVVEAFEWWSKCPRNHYPSTLYVTVRGLKVPDCIKPSYRSMMALQEEKGKPVHCERHHESSSVDTEPGLEESKEVIKWKRLPGQENEDVFVLISPTMEEY
Specifications

Specifications

AAH94800.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96.4kDa
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AHI-1/DKFZp686J1653/FLJ14023/FLJ20069/JBTS3/ORF1/dJ71N10.1
AHI1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54806
AHI1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
RUO
AHI1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications