missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AGXT Full-length ORF (BAG37731.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000189-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MASHKLLVTPLKALLKPLSIPNQLLLGPGPSNLPPRIMAAGGLQMIGSMSKDMYQIMDEIKEGIQYVFQTRNPLTLVISGSGHCALEAALVNVLEPRDSFLVGANGIWGQRAVDIGERIGARVHPMTKDPGGHYTLQEVEEGLAQHKPVLLFLTHGESSTGVLQPLDGFGELCHRYKCLLLVDSVASLGGTPLYMDRQGIDILYSGSQKALNAPPGTSLISFSDKAKKKMYSRKTKPFSFYLDIKWLANFWGCDDQPRMYHHTIPVISLYSLRESLALIAEQGLENSWRQHREAAAYLHGRLQALGLQLFVKDPALRLPTVTTVAVPAGYDWRDIVSYVMDHFDIEIMGGLGPSTGKVLRIGLLGCNATRENVDRVTEALRAALQHCPKKKLSpezifikation
BAG37731.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.52kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AGXT | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
189 | |
AGXT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGT|AGT1|AGXT1|PH1|SPAT|SPT|TLH6 | |
AGXT | |
Yes |