missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AGXT Full-length ORF (BAG37731.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
POA
Specifications
Name | AGXT (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Molecular Weight (g/mol) | 69.52kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
This gene is expressed only in the liver and the encoded protein is localized mostly in the peroxisomes, where it is involved in glyoxylate detoxification. Mutations in this gene, some of which alter subcellular targetting, have been associated with type I primary hyperoxaluria. [provided by RefSeq]
Sequence: MASHKLLVTPLKALLKPLSIPNQLLLGPGPSNLPPRIMAAGGLQMIGSMSKDMYQIMDEIKEGIQYVFQTRNPLTLVISGSGHCALEAALVNVLEPRDSFLVGANGIWGQRAVDIGERIGARVHPMTKDPGGHYTLQEVEEGLAQHKPVLLFLTHGESSTGVLQPLDGFGELCHRYKCLLLVDSVASLGGTPLYMDRQGIDILYSGSQKALNAPPGTSLISFSDKAKKKMYSRKTKPFSFYLDIKWLANFWGCDDQPRMYHHTIPVISLYSLRESLALIAEQGLENSWRQHREAAAYLHGRLQALGLQLFVKDPALRLPTVTTVAVPAGYDWRDIVSYVMDHFDIEIMGGLGPSTGKVLRIGLLGCNATRENVDRVTEALRAALQHCPKKKLSpecifications
AGXT (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
69.52kDa |