missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AGR2 Partial ORF (NP_006399, 21 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010551-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16055255

  • 545.00 € / 25µg

Estimated Shipment: 31-10-2022
to see stock.
Add to basket

Description

Description

Sequence: RDTTVKPGAKKDTKDSRPKLPQTLSRGWGDQLIWTQTYEEALYKSKTSNKPLMIIHHLDECPHSQALKKVFAENKEIQKLAEQFVLLNLV
Specifications

Specifications

NP_006399
Liquid
10551
AGR2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AG2/GOB-4/HAG-2/XAG-2
AGR2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RDTTVKPGAKKDTKDSRPKLPQTLSRGWGDQLIWTQTYEEALYKSKTSNKPLMIIHHLDECPHSQALKKVFAENKEIQKLAEQFVLLNLV
RUO
AGR2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Promotions

Promotions