missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADRBK2 Partial ORF (AAH36797, 48 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000157-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101321

  • 508.00 € / 25µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The beta-adrenergic receptor kinase specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. Overall, the beta adrenergic receptor kinase 2 has 85% amino acid similarity with beta adrenergic receptor kinase 1, with the protein kinase catalytic domain having 95% similarity. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function. [provided by RefSeq]

Sequence: AERNEITFDKIFNQKIGFLLFKDFCLNEINEAVPQVKFYEEIKEYEKLDNEEDRLCRSRQIYDAYIMKELLSCSHPFSKQAVEHVQSHLSKKQVTSTLFQ
Specifications

Specifications

AAH36797
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AERNEITFDKIFNQKIGFLLFKDFCLNEINEAVPQVKFYEEIKEYEKLDNEEDRLCRSRQIYDAYIMKELLSCSHPFSKQAVEHVQSHLSKKQVTSTLFQ
RUO
ADRBK2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
157
ADRBK2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BARK2/GRK3
ADRBK2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions