Learn More
Abnova™ Human ADCY6 Partial ORF (AAH64923, 701 a.a. - 800 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000112-Q01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes adenylate cyclase 6, which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). The expression of this gene is found in normal thyroid and brain tissues, as well as some tumors; and its expression is significantly higher in one hyperfunctioning thyroid tumor than in normal thyroid tissue. Alternative splicing generates 2 transcript variants. [provided by RefSeq]
Sequence: YASIFLLLLITVLICAVYSCGSLFPKALQRLSRSIVRSRAHSTAVGIFSVLLVFTSAIANMFTCNHTPIRSCAARMLNLTPADITACHLQQLNYSLGLDASpezifikation
AAH64923 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YASIFLLLLITVLICAVYSCGSLFPKALQRLSRSIVRSRAHSTAVGIFSVLLVFTSAIANMFTCNHTPIRSCAARMLNLTPADITACHLQQLNYSLGLDA | |
RUO | |
ADCY6 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
112 | |
ADCY6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AC6|DKFZp779F075|KIAA0422 | |
ADCY6 | |
Yes |