missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADCY6 Partial ORF (AAH64923, 701 a.a. - 800 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
POA
Specifications
Name | ADCY6 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
Molecular Weight (g/mol) | 36.63kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
This gene encodes adenylate cyclase 6, which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). The expression of this gene is found in normal thyroid and brain tissues, as well as some tumors; and its expression is significantly higher in one hyperfunctioning thyroid tumor than in normal thyroid tissue. Alternative splicing generates 2 transcript variants. [provided by RefSeq]
Sequence: YASIFLLLLITVLICAVYSCGSLFPKALQRLSRSIVRSRAHSTAVGIFSVLLVFTSAIANMFTCNHTPIRSCAARMLNLTPADITACHLQQLNYSLGLDASpecifications
ADCY6 (Human) Recombinant Protein (Q01) | |
Yes | |
36.63kDa | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |