missing translation for 'onlineSavingsMsg'
Learn More

Histone H4, Dimethyl Lys20 Antibody (1851), Novus Biologicals™

Product Code. 18149488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18149488 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18149488 Supplier Novus Biologicals Supplier No. NBP242861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence

Histone H4 (Dimethyl Lys20) Monoclonal antibody specifically detects Histone H4 (Dimethyl Lys20) in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H4 (Dimethyl Lys20)
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot
Classification Monoclonal
Clone 1851
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Dot Blot 1:500, Chromatin Immunoprecipitation (ChIP)
Formulation PBS with No Preservative
Gene Alias H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K20Me2, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Gene Symbols HIST4H4
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Histone H4K20me2 (dimethyl Lys20). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121504
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.