missing translation for 'onlineSavingsMsg'
Learn More

Heptyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Catalog Number p-7043250
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy
78617-12-6
C13H26O6
278.345
MFCD00063299
NIDYWHLDTIVRJT-UJPOAAIJSA-N
heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
448173
(2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol
CCCCCCCOC1C(C(C(C(O1)CO)O)O)O
This item is not returnable. View return policy
78617-12-6
C13H26O6
278.345
MFCD00063299
NIDYWHLDTIVRJT-UJPOAAIJSA-N
heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
448173
(2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol
CCCCCCCOC1C(C(C(C(O1)CO)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 500 mg
Beilstein 8806
Formula Weight 278.34
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Heptyl beta-D-glucopyranoside

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'