missing translation for 'onlineSavingsMsg'
Learn More

HEPES Sodium Salt (White Powder), Fisher BioReagents

Catalog Number p-26409
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
75277-39-3
C8H17N2NaO4S
260.28
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
CHEBI:46758
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
This item is not returnable. View return policy
75277-39-3
C8H17N2NaO4S
260.28
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
CHEBI:46758
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
HEPES Sodium Salt is used as a buffer for in vitro cell culture.
TRUSTED_SUSTAINABILITY
Absorbance 0.04 max. (0.1M in water, 1cm) at 260nm, 0.02 max. (0.1M in water, 1cm) at 280nm
Color White
Physical Form Powder or Solid
Quantity 1 kg
Assay Percent Range ≥99 %
Identification Pass Test
Loss on Drying 3.0% max.
Packaging Poly Bottle
Formula Weight 260.29
Percent Purity ≥99%
Chemical Name or Material HEPES Sodium Salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'