Learn More
Hepatocyte Growth Factor Ab-2 Mouse Monoclonal Antibody, Epredia™
Ideal for Western Blotting, Epredia™ Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
116.28 € - 571.20 €
Specifications
Antigen | Hepatocyte Growth Factor Ab-2 |
---|---|
Clone | SBF5 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Concentrated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12652317
|
Epredia
MS-775-P1ABX |
1mg/mL | 100 μL |
476.00 €
100µL |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12642317
|
Epredia
MS-775-P1 |
200μg/mL | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12632317
|
Epredia
MS-775-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12662317
|
Epredia
MS-775-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.Host Species: Mouse
Clone: SBF5
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human HGF protein
Molecular Weight: ∽82kDa
Recommended for:
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Hepatocyte Growth Factor Ab-2 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG1 |
SBF5 | |
Unconjugated | |
Mouse | |
Growth Factor | |
Recombinant human HGF protein | |
Primary |