Learn More
Heliotrine-N-oxide 100 μg/mL in Water, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Heliotrine-N-oxide |
| CAS | 6209-65-0 |
| Analyte or Component Names | Heliotrine-N-oxide |
| Shipping Condition | +20°C |
| Molecular Formula | C16 H27 N O6 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Butanoic acid, 2-hydroxy-2-[(1R)-1-methoxyethyl]-3-methyl-, [(1S,7aR)-2,3,5,7a-tetrahydro-1-hydroxy-4-oxido-1H-pyrrolizin-7-yl]methyl ester, (2S)-, Butanoic acid, 2-hydroxy-2-(1-methoxyethyl)-3-methyl-, (2,3,5,7a-tetrahydro-1-hydroxy-1H-pyrrolizin-7-yl)methyl ester, N-oxide, [1S-[1α,7[R*(S*)],7aα]]-, Heliotrine, 4-oxide (8CI), Heliotrine, N-oxide (6CI,7CI), (7S)-Heliotrine N-oxide, Butanoic acid, 2-hydroxy-2-(1-methoxyethyl)-3-methyl-, (2,3,5,7a-tetrahydro-1-hydroxy-4-oxido-1H-pyrrolizin-7-yl)methyl ester, [1S-[1α,7[R*(S*)],7aα]]-, Heliotrin N-oxide, Heliotrine oxide |
| InChI Formula | InChI=1S/C16H27NO6/c1-10(2)16(20,11(3)22-4)15(19)23-9-12-5-7-17(21)8-6-13(18)14(12)17/h5,10-11,13-14,18,20H,6-9H2,1-4H3/t11-,13+,14-,16+,17?/m1/s1 |
| SMILES | CO[C@H](C)[C@@](O)(C(C)C)C(=O)OCC1=CC[N+]2([O-])CC[C@H](O)[C@@H]12 |
| Show More |