missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine, MP Biomedicals™
38.15 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Use promo code "23257" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Specifications
Color | White |
Quantity | 5 g |
IUPAC Name | 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
Formula Weight | 283.2 |
Physical Form | Powder |
Chemical Name or Material | Guanosine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'