missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Graduated AC Round Bottle with Phenolic PolyCone Lined Cap
Click to view available options
No. per Case:
48/Cs.
488/Cs.
Description
- Used for storing industrial laboratory chemicals and reagents
- Designed with slight shoulder for easy pouring, handling
- Facilitates easy mixing, handling, transfer of contents
- Graduated in both milliliters and ounces
Specifications
Closure Liner | Polycone |
Closure Material | Phenolic |
Closure Type | Screw Cap |
Product Type | Graduated Bottle |
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
For Use With (Application) | Storing dry materials |
Material | Glass |
No. per Case | 48/Cs. |
Closure Color | Black |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'