missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG Fraction To Rabbit Immunoglobulins, FITC, Polyclonal Secondary Antibody, MP Biomedicals
Brand: MP Biomedicals 0855652
Additional Details : Weight : 0.00200kg
Description
Host Species: Goat
Species Reactivity: Rb
Immunofluorescence, Fluorescence Microscopy, Fluorescent Activated Cell Sorting, Western Blotting
Specifications
IgG | |
FITC | |
2 mL | |
Secondary | |
Lyophilized |
Polyclonal | |
Goat | |
RUO | |
Rabbit |