missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Pacific Orange™, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen P31584
Additional Details : Weight : 0.00450kg
Description
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry.Specifications
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Pacific Orange | |
Goat | |
purified | |
RUO | |
Rabbit | |
Liquid |
Flow Cytometry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μL | |
Secondary | |
4° C, store in dark | |
IgG |