Learn More
Fisherbrand™ Supercooled Gel Tank, Complete - For Precast Or Handcast Gels
For slab gel electrophoresis and electroblotting
Brand:  Fisherbrand™
Additional Details : Weight : 5.00000kg
Product Code. 10396432
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • A convenient and space saving solution for slab gel electrophoresis and electroblotting.
  • Combined electrophoresis and blotting - a one-tank solution for consistency of slab gel electrophoresis and electroblotting
  • Cooled vertical tank complete with casting base plus accessories, the electroblotting module and accessories
Specifications
Specifications
Supercooled Gel Tank, Complete
Supercooled gel tank, gel running module, 2 x plain glass plates with 1mm bondedspacers, 2 x notched glass plates, 2 x 1mm (24 sample) comb, 1 x dummy plate,casting base with seals and spacer aligners, internal blotting module, compressioncassettes and fibre pads (Maxi gel units 205mm x 200mm)