Learn More
Biochrom Thermostatted 5-Cell Changer
Designed to speed up analysis by allowing users to automatically measure samples that are at a controlled temperature. With a cell changer connected, the software will allow the user to perform parallel kinetics measurements on either stand-alone local control or PC-controlled instruments.
Brand: Biochrom 80-7100-09
Additional Details : Weight : 2.00000kg
Description
- For Ultrospec 7000/8000/9000
- Fully automatic operation
- Fully controlled by software, parameters stored within application methods
- Fits to standard sample compartment baseplate
- Can be changed in seconds
- Fast operation
Specifications
Ultrospec 7000/8000/9000 | |
Cell Changer | |
Measure at a Controlled Temperature | |
Ultrospec 7000/8000/9000 |