Learn More
Thermo Scientific™ Gas Guard for CO2 Tank
For Use With: 3950/3951 Incubators
Brand: Thermo Scientific™ 1900000
Additional Details : Weight : 0.10000kg
Description
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks are factory installed and built in to specified CO2 incubators; specify when ordering the incubator. Fitting unobtrusively inside the incubator casing, these systems interface directly with unit power supply and control systems.
- Factory-installed systems monitor CO2 or N2 pressure in source tanks
- Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
- Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low
Specifications
Gas Guard, CO2 | |
LED | |
Accessory, Gas Guard for CO2 Tank |