missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fritsch™ Tungsten Carbide Grinding Bowls
POA
Specifications
Product Type | Grinding Bowl |
---|---|
Composition | Tungsten carbide – TC: 93.8share%, Cobalt – Co: 6.0share%, Other: 0.2share% |
Density (Metric) | 14.95 g/cm3 |
For Use With (Application) | Hard, abrasive samples |
Hardness | 92.1HRA |
Specifications
Grinding Bowl | |
14.95 g/cm3 | |
92.1HRA | |
Tungsten Carbide |
Tungsten carbide – TC: 93.8share%, Cobalt – Co: 6.0share%, Other: 0.2share% | |
Hard, abrasive samples | |
Lid and seal ring | |
Hard, abrasive |