missing translation for 'onlineSavingsMsg'
Learn More

Fritsch™ Silicon Nitride Grinding Bowls

POA

Specifications

Product Type Grinding Bowl
Composition Al2 O3 Y2 O3: 12.5share%, Si3 N4: 86.5share%, Other: 1.00share%
Density (Metric) 3.25 g/cm3
For Use With (Application) Abrasive samples, metal-free grinding
For Use With (Equipment) Planetary Mono Mill Pulverisette 6
View More Specs

Description

Description

Specifications

Specifications

Grinding Bowl
3.25 g/cm3
Planetary Mono Mill Pulverisette 6
Lid and seal ring
Abrasive
Al2 O3 Y2 O3: 12.5share%, Si3 N4: 86.5share%, Other: 1.00share%
Abrasive samples, metal-free grinding
91.1HRA
Silicon Nitride
Videos
SDS
Documents

Documents

Product Certifications