missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOX2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40588AF405
543.15 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
FOX2 Polyclonal antibody specifically detects FOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FOX2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| dJ106I20.3, Fox-1 homolog B, fox-1 homologue, FOX2Fox-2, fxh, hexaribonucleotide binding protein 2, Hexaribonucleotide-binding protein 2, HRNBP2FOX-2, RBM9RTAHNRBP2, Repressor of tamoxifen transcriptional activity, RNA binding motif protein 9, RNA binding protein fox-1 homolog 2, RNA binding protein, fox-1 homolog (C. elegans) 2, RNA-binding motif protein 9, RNA-binding protein 9 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human RNA Binding Motif Protein 9 using the numbering given in entry NP_055124.1 (GeneID 23543). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23543 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction