Learn More
Poulten & Graf GmbH FORTUNA™ Bulb Pipette with Suction Piston
Eliminate the need for a pipetting aid. Poulten & Graf GmbH FORTUNA™ Bulb Pipette with Suction Piston ensures reliable liquid dispensing from the pipette.
15.70 € - 73.70 €
Specifications
Class | A |
---|---|
Material | Glass |
Sterility | Non-sterile |
Quantity | 1 |
Type | Bulb Pipette with suction piston |
Product Code | Brand | Volume (Metric) | Graduation Color | Tolerance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduation Color | Tolerance | Price | Quantity & Availability | |||
10588521
|
Poulten & Graf GmbH
1 2821902 |
1 mL | Blue | 0.008 mL |
15.70 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10251931
|
Poulten & Graf GmbH
1 2822502 |
2 mL | Blue | 0.010 mL |
17.50 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10572412
|
Poulten & Graf GmbH
1 2824004 |
20 mL | Amber | 0.030 mL |
29.15 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10221972
|
Poulten & Graf GmbH
1 2824104 |
25 mL | Amber | 0.03 mL |
30.10 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10202031
|
Poulten & Graf GmbH
1 2824102 |
25 mL | Blue | 0.030 mL |
30.10 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10028611
|
Poulten & Graf GmbH
1 2823104 |
5 mL | Amber | 0.015 mL |
19.65 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||
10078650
|
Poulten & Graf GmbH
1 2823102 |
5 mL | Blue | 0.015 mL |
20.40 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10100022
|
Poulten & Graf GmbH
1 2824504 |
50 mL | Amber | 0.05 mL |
73.70 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10119971
|
Poulten & Graf GmbH
1 2823604 |
10 mL | Amber | 0.020 mL |
22.55 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10159871
|
Poulten & Graf GmbH
1 2821904 |
1 mL | Amber | 0.008 mL |
15.70 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10169871
|
Poulten & Graf GmbH
1 2822504 |
2 mL | Amber | 0.010 mL |
16.85 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10483911
|
Poulten & Graf GmbH
1 2824502 |
50 mL | Blue | 0.05 mL |
73.70 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
Description
- Includes 1 mark
- Clear calibration cylinders
- No pipetting aid necessary
- Without certificate
Specifications
A | |
Non-sterile | |
Bulb Pipette with suction piston |
Glass | |
1 | |
Reusable |