missing translation for 'onlineSavingsMsg'
Learn More

FLJ20399 Recombinant Protein, Abnova

Marke:  Abnova H00054920-P01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16180893

  • 310.00 € / 10 Mikrogramm


Verfügbar ab
15-02-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spezifikation

Spezifikation

AAH06527
Solution
54920
FLJ20399 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DUS2L
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
79.97
8
Glutathione Sepharose 4 Fast Flow
10 ug
MILNSLSLCYHNKLILAPMVRVGTLPMRLLALDYGADIVYCEELIDLKMIQCKRVVNEVLSTVDFVAPDDRVVFRTCEREQNRVVFQMGTSDAERALAVARLVENDVAGIDVNMGCPKQYSTKGGMGAALLSDPDKIEKILSTLVKGTRRPVTCKIRILPSLEDTLSLVKRIERTGIAAIAVHGRKREERPQHPVSCEVIKAIADTLSIPVIANGGSHDHIQQYSDIEDFRQATAASSVMVARAAMWNPSIFLKEGLRPLEEVMQKYIRYAVQYDNHYTNTKYCLCQMLREQLESPQGRLLHAAQSSREICEAFGLGAFYEETTQELDAQQARLSAKTSEQTGEPAEDTSGVIKMAVKFDRRAYPAQITPKMCLLEWCRREKLAQPVYETVQRPLDRLFSSIVTVAEQKYQSTLWDKSKKLAEQAAAIVCLRSQGLPEGRLGEESPSLHKRKREAPDQDPGGPRAQELAQPGDLCKKPFVALGSGEESPLEGW
DUS2|FLJ20399|SMM1|URLC8
DUS2L
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation