Learn More
Fisherbrand™ Sharps-A-Gator™ Sharps Containers
Puncture-resistant with wide rectangular opening for horizontal loading of needles
POA
Specifications
Certifications/Compliance Used to comply with OSHA standards for sharps and biohazard waste collection.
Closure Color Clear
Autoclavable Autoclavable
View More Specs
Description
Description
Puncture-resistant with wide rectangular opening for horizontal loading of needles
  • General-purpose sharps container with clear locking lid and marked with biohazard label
  • Made from puncture-resistant polypropylene that can be autoclaved or incinerated
  • Easily mount on walls and carts, nest to conserve storage space
Compliance
Used to comply with OSHA standards for sharps and biohazard waste collection.
Specifications
Specifications
Used to comply with OSHA standards for sharps and biohazard waste collection.
Clear
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications