Learn More
Fisherbrand™ Standard Exhaust Oil Filters
As supplied with Fisher Scientific Maxima C Plus pumps
227.60 € - 361.23 €
Product Code | Brand | Diameter (English) Outside | Diameter (Metric) Outside | Overall Height (English) | Overall Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outside | Diameter (Metric) Outside | Overall Height (English) | Overall Height (Metric) | Price | Quantity & Availability | ||
10070011
|
Fisherbrand™
|
2-1/2 in. | 63.5 mm | 4.62 in. | 11.73 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11847044
|
Fisherbrand™
|
2-1/2 in. | 63.5 mm | 2.5 in. | 6.35 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
As supplied with Fisher Scientific Maxima C Plus pumps
These compact, round filters simply screw into threaded exhaust port to collect oil expelled during operation. A replaceable filter element within the oil filter separates the oil into particles no larger than 0.3μm, and drops collected oil back into the oil case.
- Filter element is easily replaced
- Reorder number located on filter canister
1For M4C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
2For M4C or M8C vacuum pumps designed after August 1999 (S/N FKK090000 and newer).
Specifications
Standard Exhaust Oil Filter |