missing translation for 'onlineSavingsMsg'
Learn More

Filter Aid, Celite Standard Supercel|r, Thermo Scientific Chemicals

Catalog Number 15409019
39.06 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

 

Filter Aid, Celite AW Standard Supercel™ is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter Aid, Celite AW Standard Supercel« is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals,
TRUSTED_SUSTAINABILITY
Odor Odorless
Quantity 500 g
CAS 61790-53-2
Molecular Formula O2Si
MDL Number MFCD00132803
Merck Index 14,4973
Synonym Celite 521AW, filter aid
Solubility Information Insoluble in water.
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name dioxosilane
Molecular Weight (g/mol) 60.083
PubChem CID 24261
ChEBI CHEBI:30563
Chemical Name or Material Filter Aid, Celite AW Standard Supercel™
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'