missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGFR4 Recombinant Human Protein

FGFR4 is a member of the FGFR family of receptor tyrosine kinases.

1826.00 € - 7890.00 €

Specifications

Accession Number NP_002002
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 50mM Tris, pH 7.5, 100mM NaCl, 1mM DTT, 0.01% NP-40, 0.01mM EDTA, 20% glycerol
Molecular Weight (g/mol) 42.6kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428403
View Documents Promotion Details
Invitrogen™
PR4380C
100μg
1826.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10577163
View Documents Promotion Details
Invitrogen™
PR4380A
1mg
7890.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.

Specifications

Specifications

NP_002002
50mM Tris, pH 7.5, 100mM NaCl, 1mM DTT, 0.01% NP-40, 0.01mM EDTA, 20% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
FGF Receptor Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
42.6kDa
RUO - research use only
FGFR4, JTK2
Human
His Tag
Frozen
Videos
SDS
Documents

Documents

Product Certifications