Learn More
ZVG Zellstoff Vertrieb™ FFP2 Standard Plus Filter Masks with Exhalation Valve
Fine dust FFP2 Standard Plus Filter Masks with Exhalation Valve to protect against solid and liquid particles of harmful, low-toxic substances.
Brand:  ZVG Zellstoff Vertrieb™ 3093194
Additional Details : Weight : 0.22500kg
Product Code. 10297502
  • 121.90 € / Pack of 20
Currently Unavailable: We have no information when this product will be available
View alternative products
Description
Description
  • FFP2 NR D
  • With exhalation valve
  • Easily adjusted, elastic headband
  • Hygienic packaging
  • Foam nosepad for added comfort
  • Effective against less toxic substances
Specifications
Specifications
EN 149: 2001
Single-use
Yes
Exhalation valve
Adjustable foam nosepiece
FFP2 NR D facemasks
Filter Mask
Protection Against Solid Particles of Less Toxic & Harmful Substances
Yes
20 Pk.