Learn More
ZVG Zellstoff Vertrieb™ FFP2 Standard Plus Filter Masks with Exhalation Valve
Fine dust FFP2 Standard Plus Filter Masks with Exhalation Valve to protect against solid and liquid particles of harmful, low-toxic substances.
Brand: ZVG Zellstoff Vertrieb™ 3093194
Additional Details : Weight : 0.22500kg
Description
- FFP2 NR D
- With exhalation valve
- Easily adjusted, elastic headband
- Hygienic packaging
- Foam nosepad for added comfort
- Effective against less toxic substances
Specifications
EN 149: 2001 | |
Single-use | |
Yes | |
Exhalation valve | |
Adjustable foam nosepiece |
FFP2 NR D facemasks | |
Filter Mask | |
Protection Against Solid Particles of Less Toxic & Harmful Substances | |
Yes | |
20 Pk. |