Learn More
EVOS™ Onstage Incubator Gas Line
EVOS™ Onstage Incubator Gas Line
Brand: Invitrogen AMEP4732
Additional Details : Weight : 2.00000kg
Description
The EVOS™ Onstage Incubator Gas Line connects the Control Unit and the Stagetop Environmental Chamber units of the EVOS™ Onstage Incubator and helps precisely control the CO2 and O2 levels. When performing live cell imaging studies, it is essential that the environmental conditions are precisely controlled. The EVOS™ Onstage Incubator has three gas line ports, enabling studies of cells under normal, as well as hypoxic, conditions.Explore the entire EVOS™ line of imaging systems and accessories.
Specifications
Onstage Accessory | |
EVOS™ Onstage Incubator Gas Line, Onstage Accessory, Connects the Control Unit and the Stagetop Environmental Chamber Unit, Helps Precisely Control the CO2 and O2 Levels, Use With EVOS™ FL Auto Imaging System and Cellular Imaging | |
EVOS™ FL Auto Imaging System | |
1 Each |
For Research Use Only. Not for use in diagnostic procedures.