missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%

Catalog Number p-7043113
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
60-00-4
C10H16N2O8
292.24
MFCD00003541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
6049
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
This item is not returnable. View return policy
60-00-4
C10H16N2O8
292.24
MFCD00003541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
6049
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽245°C (decomposition)
Color White
Density 0.68
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 100 g
Beilstein 1716295
Merck Index 143517
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
Formula Weight 292.24
Percent Purity ≥99.4%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'