missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 0.5M aq. soln., pH 8.0, autoclaved

Catalog Number p-7041794
39.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
60-00-4
C10H16N2O8
292.24
MFCD00003541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
6049
CHEBI:42191
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
60-00-4
C10H16N2O8
292.24
MFCD00003541
KCXVZYZYPLLWCC-UHFFFAOYSA-N
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
6049
CHEBI:42191
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Ethylenediaminetetraacetic acid is used in the treatment of lead poisoning, brain damage, cancer, rheumatoid arthritis, osteoarthritis, an eye condition called macular degeneration, diabetes, Alzheimer's disease, multiple sclerosis and Parkinson's disease. It acts as a chelating agent and used for the preparation of many solutions including DNA loading dye and resuspension buffer which is isolation of plasmid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and copper.
TRUSTED_SUSTAINABILITY
pH 8.0
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 250 mL
Solubility Information Miscible with water.
Formula Weight 292.24
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Physical Form Liquid
Chemical Name or Material Ethylenediaminetetraacetic acid, autoclaved

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'