missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium sa, Thermo Scientific Chemicals

Catalog Number p-4112115
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
13368-13-3
C14H20N2Na4O10
468.28
MFCD00274420
BAOGCDPNNOBWQZ-UHFFFAOYSA-J
egta tetrasodium, 3,12-bis carboxymethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, ethylene glycol-bis beta-aminoethyl ether-n,n,n',n'-tetraacetic acid tetrasodium salt, egta tetrasodium salt, egtazic acid tetrasodium, ethylene-bis oxyethylenenitrilo tetraacetic acid tetrasodium, 2,2',2,2'-ethylenebis oxyethylenenitrilo tetraacetic acid tetrasodium salt, 6,9-dioxa-3,12-diazatetradecanedioicacid,3,12-bis carboxymethyl-,sodium salt 1:4, tetrasodium 3,12-bis carboxylatomethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioate, ethylene glycol-bis, a-aminoethyl ether-n,n,n inverted exclamation marka,n inverted exclamation marka-tetraacetic acid tetrasodium salt
16219327
tetrasodium;2-[2-[2-[2-[bis(carboxylatomethyl)amino]ethoxy]ethoxy]ethyl-(carboxylatomethyl)amino]acetate
C(COCCOCCN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+].[Na+].[Na+]
13368-13-3
C14H20N2Na4O10
468.28
MFCD00274420
BAOGCDPNNOBWQZ-UHFFFAOYSA-J
egta tetrasodium, 3,12-bis carboxymethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, ethylene glycol-bis beta-aminoethyl ether-n,n,n',n'-tetraacetic acid tetrasodium salt, egta tetrasodium salt, egtazic acid tetrasodium, ethylene-bis oxyethylenenitrilo tetraacetic acid tetrasodium, 2,2',2,2'-ethylenebis oxyethylenenitrilo tetraacetic acid tetrasodium salt, 6,9-dioxa-3,12-diazatetradecanedioicacid,3,12-bis carboxymethyl-,sodium salt 1:4, tetrasodium 3,12-bis carboxylatomethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioate, ethylene glycol-bis, a-aminoethyl ether-n,n,n inverted exclamation marka,n inverted exclamation marka-tetraacetic acid tetrasodium salt
16219327
tetrasodium;2-[2-[2-[2-[bis(carboxylatomethyl)amino]ethoxy]ethoxy]ethyl-(carboxylatomethyl)amino]acetate
C(COCCOCCN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 54.0°C to 56.0°C
Assay Percent Range 18.46 to 20.82% (ICP on dried sample)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble.
Formula Weight 468.28
Percent Purity 95%
Chemical Name or Material Ethylene glycol-bis(β-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt
compliance-icons
Product Identifier
  • Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'