missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (trimethylsilyl)acetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7023800
26.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4071-88-9
C7H16O2Si
160.288
MFCD00009172
QQFBQBDINHJDMN-UHFFFAOYSA-N
ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
77687
ethyl 2-trimethylsilylacetate
CCOC(=O)C[Si](C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4071-88-9
C7H16O2Si
160.288
MFCD00009172
QQFBQBDINHJDMN-UHFFFAOYSA-N
ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
77687
ethyl 2-trimethylsilylacetate
CCOC(=O)C[Si](C)(C)C

Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 0.876
Boiling Point 156°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.415
Quantity 10 g
UN Number UN3272
Beilstein 1755902
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 160.29
Percent Purity ≥98%
Chemical Name or Material Ethyl (trimethylsilyl)acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'