missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methoxyacetate, 98%

Product Code. p-7024326
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11453048

Brand: Thermo Scientific Alfa Aesar A18770.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3938-96-3 | C5H10O3 | 118.132 g/mol

Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3938-96-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00009176
InChI Key JLEKJZUYWFJPMB-UHFFFAOYSA-N
Synonym ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
PubChem CID 77544
IUPAC Name ethyl 2-methoxyacetate
SMILES CCOC(=O)COC

Specifications

Boiling Point 44°C to 46°C (9 mmHg)
Flash Point 46°C (115°F)
Quantity 10 g
UN Number UN3272
Beilstein 1744761
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Ethyl methoxyacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.