Learn More
Ethyl diacetoacetate, 97%
Product Code. 11430691
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11430691
Brand: Thermo Scientific Alfa Aesar B20149.09
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 603-69-0 | C8H12O4 | 172.18 g/mol
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl diacetoacetate is used in the preparation of 3-acetyl-6-chloro-7-hydroxy-4-methylcoumarin. It is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 603-69-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009864
InChI Key YMCDYRGMTRCAPZ-UHFFFAOYSA-N
PubChem CID 79063
IUPAC Name ethyl 2-acetyl-3-oxobutanoate
SMILES CCOC(=O)C(C(=O)C)C(=O)C
Specifications
Density 1.104
Boiling Point 104°C to 105°C (17 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.472
Quantity 10 g
Beilstein 775014
Synonym ethyl diacetoacetate, ethyl 2-acetylacetoacetate, butanoic acid, 2-acetyl-3-oxo-, ethyl ester, 3-ethoxycarbonylacetylacetone, ethyl diacetylacetate, ethyl diacetyl acetate, ethyl diaceto acetate, 2-acetyl-3-oxo-butyric acid ethyl ester, butanoic acid,2-acetyl-3-oxo-, ethyl ester, ethyldiacetoacetate
Solubility Information Not miscible in water.
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Ethyl diacetoacetate