missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromo-2-methylnicotinate, 97%, Thermo Scientific™

Catalog Number 15427918
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
129477-21-0
C9H10BrNO2
244.088
MFCD10698571
QVKPPFGWQMCQCI-UHFFFAOYSA-N
15077476
ethyl 5-bromo-2-methylpyridine-3-carboxylate
CCOC(=O)C1=CC(=CN=C1C)Br
This item is not returnable. View return policy
129477-21-0
C9H10BrNO2
244.088
MFCD10698571
QVKPPFGWQMCQCI-UHFFFAOYSA-N
15077476
ethyl 5-bromo-2-methylpyridine-3-carboxylate
CCOC(=O)C1=CC(=CN=C1C)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 5-bromo-2-methylnicotinate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Synonym ethyl 5-bromo-2-methylnicotinate, 5-bromo-2-methyl-nicotinic acid ethyl ester, 5-bromo-2-methyl-3-pyridinecarboxylic acid ethyl ester, 5-bromo-2-methylnicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-methyl-, ethyl ester, ethyl5-bromo-2-methylnicotinate, ethyl 5-bromo-2-methyl-pyridine-3-carboxylate
Solubility Information Slightly soluble in water.
Formula Weight 244.09
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 5-bromo-2-methylnicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'